1,2-Epoxy-3-methylbutane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19291-03
L19291-06
CAAAAL19291-03EA
106.49
CAD
CAAAAL19291-03
CAAAAL19291-06
1,2-Epoxy-3-methylbutane 98+%
1,2-Epoxy-3-methylbutane
Purity: 98+%
CAS No.: 1438-14-8
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: -12°(10°F)
Refractive Index: 1.3915
Odor: Recognizable
Boiling Point: 72-75°
Beilstein Reference: 102418
UN No.: 1993
MDL No.: MFCD02683501
Formula:
C₅H₁₀O Boiling Pt: 72…75 °C Density: 0.815 Flash Pt: –12 °C (10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02683501 CAS Number: 1438-14-8 EINECS: 215-869-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...