6-Aminonicotinonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19282-06
L19282-14
CAAAAL19282-06EA
57.2
CAD
CAAAAL19282-06
CAAAAL19282-14
6-Aminonicotinonitrile 98%
6-Aminonicotinonitrile
Purity: 98%
CAS No.: 4214-73-7
Molecular Formula: C6H5N3
Formula Weight: 119.13
Melting Point: 159-164°
Boiling Point: 240-250°/15mm
UN No.: 3439
MDL No.: MFCD00128902
Formula:
C₆H₅N₃ Boiling Pt: 240…250 °C/15 mm Melting Pt: 159…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128902 CAS Number: 4214-73-7 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...