1-Boc-3-piperidone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-BOC-3-piperidone, 1-Boc-3-Piperidinone
L19276-03
L19276-06
CAAAAL19276-03EA
35.55
CAD
CAAAAL19276-03
CAAAAL19276-06
1-Boc-3-piperidone 97%
1-Boc-3-piperidone
Purity: 97%
CAS No.: 98977-36-7
Molecular Formula: C10H17NO3
Formula Weight: 199.25
Melting Point: 37-39°
Beilstein Reference: 5936353
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol Melting Pt: 37…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631193 CAS Number: 98977-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...