3-Bromo-4-chlorobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19244-03
L19244-06
CAAAAL19244-03EA
87.69
CAD
CAAAAL19244-03
CAAAAL19244-06
3-Bromo-4-chlorobenzoic acid 97%
3-Bromo-4-chlorobenzoic acid
Purity: 97%
CAS No.: 42860-10-6
Molecular Formula: C7H4BrClO2
Formula Weight: 235.47
Melting Point: 218-222°
Beilstein Reference: 2438796
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol Melting Pt: 218…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079706 CAS Number: 42860-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...