2-Fluoro-5-nitrobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19233-03
L19233-06
CAAAAL19233-03EA
60.2
CAD
CAAAAL19233-03
CAAAAL19233-06
2-Fluoro-5-nitrobenzaldehyde 98%
2-Fluoro-5-nitrobenzaldehyde
Purity: 98%
CAS No.: 27996-87-8
Molecular Formula: C7H4FNO3
Formula Weight: 169.11
Melting Point: 57-59°
Beilstein Reference: 1950329
Formula:
C₇H₄FNO₃ Melting Pt: 57…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042298 CAS Number: 27996-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...