(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid
L19153-03
L19153-06
CAAAAL19153-03EA
107.12
CAD
CAAAAL19153-03
CAAAAL19153-06
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid 97%
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid
Purity: 97%
CAS No.: 22893-39-6
Molecular Formula: C9H6ClF3O2
Formula Weight: 238.59
Melting Point: 112-114°
Beilstein Reference: 2053702
Formula:
C₉H₆ClF₃O₂ MW: 238.59 g/mol Melting Pt: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631471 CAS Number: 22893-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...