(S)-(-)-α-Methylbenzylamine ≥99%, ee 99.5%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19118-06
L19118-14
L19118-22
CAAAAL19118-06EA
54.35
CAD
CAAAAL19118-06
CAAAAL19118-14
CAAAAL19118-22
(S)-(-)-α-Methylbenzylamine ≥99%, ee 99.5%, ChiPros®
(S)-(-)-α-Methylbenzylamine
Purity: 99+%
CAS No.: 2627-86-3
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 71°(159°F)
Refractive Index: 1.5260
Odor: Amine-like
Melting Point: -10°
Boiling Point: 187°
Beilstein Reference: 2204907
Merck Reference: 14,6026
Optical Rotation: -30° (c=10 in ethanol)
UN No.: 2735
MDL No.: MFCD00064406
Formula:
C₈H₁₁N Boiling Pt: 187 °C Melting Pt: –10 °C Density: 0.948 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064406 CAS Number: 2627-86-3 EINECS: 220-098-0 UN: 2735 ADR: 8,III Merck Index: 14,6026 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...