(R)-(+)-α-Methylbenzylamine ≥99%, ee 99%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
L19117-06
L19117-14
L19117-22
CAAAAL19117-06EA
59.41
CAD
CAAAAL19117-06
CAAAAL19117-14
CAAAAL19117-22
(R)-(+)-α-Methylbenzylamine ≥99%, ee 99%, ChiPros®
(R)-(+)-α-Methylbenzylamine
Purity: 99+%
CAS No.: 3886-69-9
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 71°(159°F)
Refractive Index: 1.5260
Odor: Amine-like
Melting Point: -10°
Boiling Point: 187°
Beilstein Reference: 2410916
Merck Reference: 14,6026
Optical Rotation: +30° (c=10 in ethanol)
UN No.: 2735
MDL No.: MFCD00064405
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 187 °C Melting Pt: –10 °C Density: 0.948 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064405 CAS Number: 3886-69-9 EINECS: 223-423-4 UN: 2922 ADR: 8 III Merck Index: 14,6026 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...