1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]

L19087-09 L19087-18
CAAAAL19087-09EA 181.7 CAD
CAAAAL19087-09 CAAAAL19087-18
1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%
1-Butyl-3-methylimidazolium tetrafluoroborate

Purity: 98+% CAS No.: 174501-65-6 Molecular Formula: C8H15BF4N2 Formula Weight: 226.03 Flash Point: None Refractive Index: 1.4220 MDL No.: MFCD03095449

Formula: C₈H₁₅BF₄N₂
MW: 226.02 g/mol
Boiling Pt: ? 200 °C (1013 hPa)
Melting Pt: –71 °C
Density: 1.210
Flash Pt: ≥ 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03095449
CAS Number: 174501-65-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR