1-Butyl-3-methylimidazoliumhexafluorophosphate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V), 1-(But-1-yl)-3-methyl-1H-imidazol-3-ium hexafluorophosphate

L19086-06 L19086-14 L19086-22
CAAAAL19086-06EA 71.26 CAD
CAAAAL19086-06 CAAAAL19086-14 CAAAAL19086-22
1-Butyl-3-methylimidazoliumhexafluorophosphate 98+%
1-Butyl-3-methylimidazolium hexafluorophosphate

Purity: 98+% CAS No.: 174501-64-5 Molecular Formula: C8H15F6N2P Formula Weight: 284.19 Refractive Index: 1.4115 Melting Point: 8-10° Boiling Point: 350° Merck Reference: 14,1581 MDL No.: MFCD03093295

Formula: C₈H₁₅F₆N₂P
Boiling Pt: >350 °C
Melting Pt: 8…10 °C
Density: 1.365
Flash Pt: ≥ 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03093295
CAS Number: 174501-64-5
Merck Index: 14,1581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR