1-Bromo-4-ethynylbenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne

L19082-03 L19082-MD
CAAAAL19082-03EA 145.99 CAD
CAAAAL19082-03 CAAAAL19082-MD
1-Bromo-4-ethynylbenzene 97%
1-Bromo-4-ethynylbenzene

Purity: 97% CAS No.: 766-96-1 Molecular Formula: C8H5Br Formula Weight: 181.04 Odor: Odorless Melting Point: 64-67° Boiling Point: 88-90°/16mm Beilstein Reference: 1209726 MDL No.: MFCD00168822

Formula: C₈H₅Br
MW: 181.03 g/mol
Boiling Pt: 88…90 °C/16 mm
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00168822
CAS Number: 766-96-1
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR