5-Methoxyindol-3-ylacetic acid 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-Methoxy-3-indoleacetic acid, 2-(5-Methoxy-1H-indol-3-yl)acetic acid, 5-Methoxyindole-3-acetic acid
L19073-03
L19073-MD
CAAAAL19073-03EA
211.72
CAD
CAAAAL19073-03
CAAAAL19073-MD
5-Methoxyindol-3-ylacetic acid 98+%
5-Methoxyindol-3-ylacetic acid
Purity: 98+%
CAS No.: 3471-31-6
Molecular Formula: C11H11NO3
Formula Weight: 205.22
Melting Point: 145-148° dec.
Beilstein Reference: 187161
Formula:
C₁₁H₁₁NO₃ MW: 205.21 g/mol Melting Pt: 145…148 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005638 CAS Number: 3471-31-6 EINECS: 222-438-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...