3-Hydroxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19061-03
L19061-06
CAAAAL19061-03EA
70.15
CAD
CAAAAL19061-03
CAAAAL19061-06
3-Hydroxyphenylboronic acid 97%
3-Hydroxyphenylboronic acid
Purity: 97%
CAS No.: 87199-18-6
Molecular Formula: C6H7BO3
Formula Weight: 137.93
Melting Point: 208-210°
Beilstein Reference: 3240771
Formula:
C₆H₇BO₃ Melting Pt: 208…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074603 CAS Number: 87199-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...