o-Toluenesulfonyl chloride 80%

Supplier: Thermo Scientific Chemicals
Danger

L18983-06 L18983-14
CAAAAL18983-06EA 131.77 CAD
CAAAAL18983-06 CAAAAL18983-14
o-Toluenesulfonyl chloride 80%
o-Toluenesulphonyl chloride

Purity: 97% CAS No.: 133-59-5 Molecular Formula: C7H7ClO2S Formula Weight: 190.65 Flash Point: 110°(230°F) Refractive Index: 1.5560 Melting Point: 10° Boiling Point: 154°/10mm Beilstein Reference: 973180 UN No.: 3265 MDL No.: MFCD00024874

Formula: C₇H₇ClO₂S
Boiling Pt: 154 °C/10 mm
Melting Pt: 10 °C
Density: 1.338
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00024874
CAS Number: 133-59-5
EINECS: 205-113-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR