Methyl nitroacetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18950-04
L18950-09
CAAAAL18950-04EA
127.03
CAD
CAAAAL18950-04
CAAAAL18950-09
Methyl nitroacetate 98%
Methyl nitroacetate
Purity: 98%
CAS No.: 2483-57-0
Molecular Formula: C3H5NO4
Formula Weight: 119.08
Flash Point: 105°(221°F)
Refractive Index: 1.4260
Odor: Ester-like
Boiling Point: 195-198°
Beilstein Reference: 1758670
MDL No.: MFCD00075480
Formula:
C₃H₅NO₄ Boiling Pt: 195…198 °C Density: 1.294 Flash Pt: 105 °C(221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075480 CAS Number: 2483-57-0 EINECS: 219-622-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...