2-(2-Aminoethoxy)ethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

L18897-22 L18897-36
CAAAAL18897-22EA 68.89 CAD
CAAAAL18897-22 CAAAAL18897-36
2-(2-Aminoethoxy)ethanol 98%
2-(2-Aminoethoxy)ethanol

Purity: 98% CAS No.: 929-06-6 Molecular Formula: H2NCH2CH2OCH2CH2OH Formula Weight: 105.14 Flash Point: 127°(260°F) Refractive Index: 1.4570 Melting Point: -11° Boiling Point: 125-131°/23mm Beilstein Reference: 906728 UN No.: 3055 MDL No.: MFCD00008181

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 220
Melting Pt: -11
Density: 1.057
Flash Pt: 127 °C (260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008181
CAS Number: 929-06-6
EINECS: 213-195-4
UN: 3055
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR