1-Boc-2-piperidone 99%
Supplier: Thermo Scientific Chemicals
L18896-03
L18896-06
CAAAAL18896-03EA
22.28
CAD
CAAAAL18896-03
CAAAAL18896-06
1-Boc-2-piperidone 99%
1-Boc-2-piperidone
Purity: 99%
CAS No.: 85908-96-9
Molecular Formula: C10H17NO3
Formula Weight: 199.25
Melting Point: 34-37°
Beilstein Reference: 4674955
Formula:
C₁₀H₁₇NO₃ Melting Pt: 34…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179046 CAS Number: 85908-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...