(S)-(+)-6-Methyl-5-hepten-2-ol 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L18884-03
L18884-MD
CAAAAL18884-03EA
262.28
CAD
CAAAAL18884-03
CAAAAL18884-MD
(S)-(+)-6-Methyl-5-hepten-2-ol 99%
(S)-(+)-6-Methyl-5-hepten-2-ol
Purity: 99%
CAS No.: 58917-26-3
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 67°(152°F)
Refractive Index: 1.4480
Boiling Point: 78-80°/15mm
Beilstein Reference: 1720071
Optical Rotation: +18° (neat)
MDL No.: MFCD03093080
Formula:
C₈H₁₆O Boiling Pt: 78…80 °C/15 mm Density: 0.844 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03093080 CAS Number: 58917-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...