(S)-(+)-4-Methyl-2-pentanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18882-03
L18882-MD
CAAAAL18882-03EA
319.16
CAD
CAAAAL18882-03
CAAAAL18882-MD
(S)-(+)-4-Methyl-2-pentanol 99%
(S)-(+)-4-Methyl-2-pentanol
For synthesis of optically active products
Purity: 99%
CAS No.: 14898-80-7
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 41°(105°F)
Refractive Index: 1.4110
Odor: Slightly sweetish
Boiling Point: 130-131°
Beilstein Reference: 1718991
Optical Rotation: +21° (neat)
UN No.: 2053
MDL No.: MFCD03093078
Formula:
C₆H₁₄O Boiling Pt: 130…131 °C Density: 0.802 Flash Pt: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03093078 CAS Number: 14898-80-7 UN: 2053 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...