(R)-(+)-3-Butyne-2-ol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne
L18880-03
L18880-06
L18880-MD
CAAAAL18880-03EA
230.68
CAD
CAAAAL18880-03
CAAAAL18880-06
CAAAAL18880-MD
(R)-(+)-3-Butyne-2-ol 99%
(R)-(+)-3-Butyne-2-ol
Purity: 99%
CAS No.: 42969-65-3
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 28°(82°F)
Refractive Index: 1.4260
Boiling Point: 108-109°
Beilstein Reference: 3587272
Optical Rotation: +47° (neat)
UN No.: 2929
MDL No.: MFCD00211237
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 103 Density: 0.891 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00211237 CAS Number: 42969-65-3 UN: 2929 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...