m-Ethyltoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

L18788-06 L18788-14
CAAAAL18788-06EA 103.17 CAD
CAAAAL18788-06 CAAAAL18788-14
m-Ethyltoluene 97%
m-Ethyltoluene

Purity: 97% CAS No.: 620-14-4 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 38°(100°F) Refractive Index: 1.4960 Melting Point: -96° Boiling Point: 158-159° Beilstein Reference: 1850821 UN No.: 3295 MDL No.: MFCD00009259

Formula: C₉H₁₂
Boiling Pt: 158…159 °C
Melting Pt: –96 °C
Density: 0.865
Flash Pt: 38 °C (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009259
CAS Number: 620-14-4
EINECS: 210-626-8
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR