1,2-Diethylbenzene 97%

Supplier: Thermo Scientific Chemicals

L18775-04 L18775-09 L18775-18
CAAAAL18775-04EA 98.91 CAD
CAAAAL18775-04 CAAAAL18775-09 CAAAAL18775-18
1,2-Diethylbenzene 97%
1,2-Diethylbenzene

Purity: 97% CAS No.: 135-01-3 Molecular Formula: C10H14 Formula Weight: 134.22 Flash Point: 55°(131°F) Refractive Index: 1.5050 Melting Point: -31° Boiling Point: 183° Beilstein Reference: 1904392 UN No.: 2049 MDL No.: MFCD00009258

Formula: C₁₀H₁₄
Boiling Pt: 183 °C
Melting Pt: –31 °C
Density: 0.880
MDL Number: MFCD00009258
CAS Number: 135-01-3
EINECS: 205-170-1
UN: 2049
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR