Allyl isocyanate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18710-03
L18710-06
CAAAAL18710-03EA
62.73
CAD
CAAAAL18710-03
CAAAAL18710-06
Allyl isocyanate 96%
Allyl isocyanate
Purity: 96%
CAS No.: 1476-23-9
Molecular Formula: CH2=CHCH2NCO
Formula Weight: 83.09
Storage Temperature: 2°C to 8°C
Flash Point: 6°(43°F)
Refractive Index: 1.4185
Odor: Pungent
Boiling Point: 87-89°
Beilstein Reference: 506106
UN No.: 2478
MDL No.: MFCD00002044
Formula:
C₄H₅NO Boiling Pt: 87…89 °C Density: 0.940 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002044 CAS Number: 1476-23-9 UN: 2478 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...