1-(3-Hydroxyphenyl)piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
m-(1-Piperazinyl)phenol, 3-(1-Piperazinyl)phenol
L18702-06
L18702-14
CAAAAL18702-06EA
60.83
CAD
CAAAAL18702-06
CAAAAL18702-14
1-(3-Hydroxyphenyl)piperazine 97%
1-(3-Hydroxyphenyl)piperazine
Purity: 98%
CAS No.: 59817-32-2
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 205-208°
Beilstein Reference: 4248583
UN No.: 3263
MDL No.: MFCD00052896
Formula:
C₁₀H₁₄N₂O MW: 178.23 Melting Pt: 205…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052896 CAS Number: 59817-32-2 UN: 3263 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...