1-(4-Nitrophenyl)piperazine 98+%

Supplier: Thermo Scientific Chemicals
Warning

L18700-14 L18700-22
CAAAAL18700-14EA 152.63 CAD
CAAAAL18700-14 CAAAAL18700-22
1-(4-Nitrophenyl)piperazine 98+%
1-(4-Nitrophenyl)piperazine

Purity: 98+% CAS No.: 6269-89-2 Molecular Formula: C10H13N3O2 Formula Weight: 207.23 Melting Point: 129-133° UN No.: 2811 MDL No.: MFCD00005961

Formula: C₁₀H₁₃N₃O₂
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005961
CAS Number: 6269-89-2
EINECS: 228-443-7
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR