4-(Trifluoromethoxy)thiophenol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Trifluoromethoxy)benzenethiol, 4-(Trifluoromethoxy)benzene-1-thiol

L18674-03 L18674-06
CAAAAL18674-03EA 107.76 CAD
CAAAAL18674-03 CAAAAL18674-06
4-(Trifluoromethoxy)thiophenol 95%
4-(Trifluoromethoxy)thiophenol

Purity: 95% CAS No.: 169685-29-4 Molecular Formula: C7H5F3OS Formula Weight: 194.18 Refractive Index: 1.4750 Boiling Point: 64-66°/15mm UN No.: 2810 MDL No.: MFCD00052312

Formula: C₇H₅F₃OS
MW: 194.18 g/mol
Boiling Pt: 64…66 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00052312
CAS Number: 169685-29-4
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR