3,5-Dimethylbenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L18603-03
L18603-06
CAAAAL18603-03EA
50.24
CAD
CAAAAL18603-03
CAAAAL18603-06
3,5-Dimethylbenzenesulfonyl chloride 97%
3,5-Dimethylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 2905-27-3
Molecular Formula: C8H9ClO2S
Formula Weight: 204.68
Melting Point: 92-94°
Beilstein Reference: 2640617
UN No.: 3261
MDL No.: MFCD03094655
Formula:
C₈H₉ClO₂S Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094655 CAS Number: 2905-27-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...