2-Bromophenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18581-03
L18581-06
CAAAAL18581-03EA
65.89
CAD
CAAAAL18581-03
CAAAAL18581-06
2-Bromophenylboronic acid 98%
2-Bromophenylboronic acid
Purity: 98%
CAS No.: 244205-40-1
Molecular Formula: C6H6BBrO2
Formula Weight: 200.83
Melting Point: 108-110°
Beilstein Reference: 8542615
Formula:
C₆H₆BBrO₂ Melting Pt: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01114672 CAS Number: 244205-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...