(R)-(-)-3-Chloro-1,2-propanediol 97%, ee 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(-)-3-Chloro-1,2-dihydroxy propane, (2R)-(-)-3-Chloropropane-1,2-diol
L18555-06
L18555-14
CAAAAL18555-03EA
63.58
CAD
CAAAAL18555-03
CAAAAL18555-06
CAAAAL18555-14
(R)-(-)-3-Chloro-1,2-propanediol 97%, ee 98%
(R)-(-)-3-Chloro-1,2-propanediol
Purity: 97%
CAS No.: 57090-45-6
Molecular Formula: C3H7ClO2
Formula Weight: 110.54
Flash Point: 137°(279°F)
Refractive Index: 1.4800
Boiling Point: 213°
Beilstein Reference: 2202928
Optical Rotation: -0.9°(neat)
UN No.: 2689
MDL No.: MFCD00135169
Formula:
ClCH₂CHOHCH₂OH Boiling Pt: 213 °C Density: 1.322 Flash Pt: 137 °C (279 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00135169 CAS Number: 57090-45-6 EINECS: 435-560-9 UN: 2689 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...