3,4,5-Trifluorophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18519-03
L18519-MD
CAAAAL18519-03EA
295.46
CAD
CAAAAL18519-03
CAAAAL18519-MD
3,4,5-Trifluorophenylboronic acid 97%
3,4,5-Trifluorophenylboronic acid
Purity: 97%
CAS No.: 143418-49-9
Molecular Formula: C6H4BF3O2
Formula Weight: 175.90
Melting Point: 275-279°
Beilstein Reference: 7371914
Formula:
C₆H₄BF₃O₂ Melting Pt: 275…279 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093069 CAS Number: 143418-49-9 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...