Aluminium chloride, anhydrous ≥99%, powder

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum chloride

L18489-22 L18489-36
CAAAAL18489-22EA 86.43 CAD
CAAAAL18489-22 CAAAAL18489-36
Aluminium chloride, anhydrous ≥99%, powder
Aluminium chloride

Purity: 99% CAS No.: 7446-70-0 Molecular Formula: AlCl3 Formula Weight: 133.34 Odor: Pungent pH: 2,4 Melting Point: 190° subl Merck Reference: 14,337 UN No.: 1726 MDL No.: MFCD00003422

Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 190 °C (subl.)
Density: 2.44
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II
Merck Index: 14,337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR