2,4-Dichloro-5-isopropoxyaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

L18371-03 L18371-06
CAAAAL18371-03EA 118.34 CAD
CAAAAL18371-03 CAAAAL18371-06
2,4-Dichloro-5-isopropoxyaniline 98%
2,4-Dichloro-5-isopropoxyaniline

Purity: 98% CAS No.: 41200-96-8 Molecular Formula: C9H11Cl2NO Formula Weight: 220.11 Melting Point: 40-44° Boiling Point: 110-112°/0.2mm UN No.: 2811 MDL No.: MFCD00974411

Formula: C₉H₁₁Cl₂NO
Boiling Pt: 110…112 °C/0.2 mm
Melting Pt: 40…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00974411
CAS Number: 41200-96-8
EINECS: 255-258-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR