6-Chloro-o-toluic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18348-03
L18348-06
CAAAAL18348-03EA
111.71
CAD
CAAAAL18348-03
CAAAAL18348-06
6-Chloro-o-toluic acid 97%
6-Chloro-o-toluic acid
Purity: 97%
CAS No.: 21327-86-6
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Melting Point: 103-105°
Beilstein Reference: 2614092
Formula:
C₈H₇ClO₂ Melting Pt: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045799 CAS Number: 21327-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...