Oxazole 98+%

Supplier: Thermo Scientific Chemicals
Danger

L18341-03 L18341-06
CAAAAL18341-03EA 186.44 CAD
CAAAAL18341-03 CAAAAL18341-06
Oxazole 98+%
Oxazole

Purity: 98+% CAS No.: 288-42-6 Molecular Formula: C3H3NO Formula Weight: 69.07 Storage Temperature: 2°C to 8°C Flash Point: 19°(66°F) Refractive Index: 1.4270 Melting Point: -86° Boiling Point: 69-70° Beilstein Reference: 103851 UN No.: 2924 MDL No.: MFCD00009751

Formula: C₃H₃NO
MW: 69.06 g/mol
Boiling Pt: 69…70 °C
Melting Pt: –86 °C
Density: 1.050
Flash Pt: 19 °C (66 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009751
CAS Number: 288-42-6
EINECS: 206-020-8
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR