3-(3-Bromophenyl)propionic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L18080-03 L18080-06
CAAAAL18080-03EA 46.45 CAD
CAAAAL18080-03 CAAAAL18080-06
3-(3-Bromophenyl)propionic acid 97%
3-(3-Bromophenyl)propionic acid

Purity: 97% CAS No.: 42287-90-1 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Melting Point: 63-65° Beilstein Reference: 2362753

Formula: C₉H₉BrO₂
MW: 229.07
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01310792
CAS Number: 42287-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR