2-Aminophenylboronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L18069-03
L18069-06
CAAAAL18069-03EA
281.24
CAD
CAAAAL18069-03
CAAAAL18069-06
CAAAAL18069-MC
2-Aminophenylboronic acid 96%
2-Aminophenylboronic acid
Purity: 96%
CAS No.: 5570-18-3
Molecular Formula: C6H8BNO2
Formula Weight: 136.95
Melting Point: 188-190°
Beilstein Reference: 2962939
Formula:
C₆H₈BNO₂ Melting Pt: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074645 CAS Number: 5570-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...