1-Boc-indole-2-boronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
L18009-03
L18009-06
L18009-MD
CAAAAL18009-03EA
169.06
CAD
CAAAAL18009-03
CAAAAL18009-06
CAAAAL18009-MD
1-Boc-indole-2-boronic acid 95%
1-Boc-indole-2-boronic acid
Purity: 95%
CAS No.: 213318-44-6
Molecular Formula: C13H16BNO4
Formula Weight: 261.08
Storage Temperature: 2°C to 8°C
Melting Point: ca 85° dec.
Beilstein Reference: 8148683
MDL No.: MFCD02093045
Formula:
C₁₃H₁₆BNO₄ MW: 261.08 g/mol Melting Pt: ca 85 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02093045 CAS Number: 213318-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...