4-Cyanophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

L18007-03 L18007-06
CAAAAL18007-03EA 93.22 CAD
CAAAAL18007-03 CAAAAL18007-06
4-Cyanophenylboronic acid 98%
4-Cyanophenylboronic acid

Purity: 98% CAS No.: 126747-14-6 Molecular Formula: C7H6BNO2 Formula Weight: 146.94 Melting Point: 300° Beilstein Reference: 6593772

Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318968
CAS Number: 126747-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR