Potassium (3-bromophenyl)trifluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
potassium (3-bromophenyl)trifluoroborate, (3-Bromophenyl)trifluoroboric acid potassium salt
L17966-03
L17966-06
CAAAAL17966-03EA
41.24
CAD
CAAAAL17966-03
CAAAAL17966-06
Potassium (3-bromophenyl)trifluoroborate 97%
Potassium (3-bromophenyl)trifluoroborate
Purity: 97%
CAS No.: 374564-34-8
Molecular Formula: C6H4BBrF3K
Formula Weight: 262.91
Melting Point: 178-182°
MDL No.: MFCD02093977
Formula:
C₆H₄BBrF₃K MW: 262.91 g/mol Melting Pt: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093977 CAS Number: 374564-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...