4-Amino-3-methylbenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17868-03
L17868-06
CAAAAL17868-03EA
41.08
CAD
CAAAAL17868-03
CAAAAL17868-06
4-Amino-3-methylbenzonitrile 98%
4-Amino-3-methylbenzonitrile
Purity: 98%
CAS No.: 78881-21-7
Molecular Formula: C8H8N2
Formula Weight: 132.17
Melting Point: 93-94°
Beilstein Reference: 3235728
UN No.: 3439
MDL No.: MFCD02093969
Formula:
C₈H₈N₂ Boiling Pt: 292 °C (1013 hPa) Melting Pt: 93…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093969 CAS Number: 78881-21-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...