3,4-Difluorobenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,4-Difluorobenzene-1-sulfonyl chloride, 3,4-Difluorobenzene-1-sulphonyl chloride
L17816-03
L17816-06
CAAAAL17816-03EA
73.79
CAD
CAAAAL17816-03
CAAAAL17816-06
3,4-Difluorobenzenesulfonyl chloride 97%
3,4-Difluorobenzenesulphonyl chloride
Purity: 97%
CAS No.: 145758-05-0
Molecular Formula: C6H3ClF2O2S
Formula Weight: 212.60
Refractive Index: 1.5140
Boiling Point: 105°/15mm
UN No.: 3265
MDL No.: MFCD00153120
Formula:
C₆H₃ClF₂O₂S MW: 212.6 g/mol Boiling Pt: 105 °C/15 mm Density: 1.586 Storage Temperature: Ambient |
MDL Number:
MFCD00153120 CAS Number: 145758-05-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...