3-Iodobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

L17801-03 L17801-06
CAAAAL17801-03EA 172.22 CAD
CAAAAL17801-03 CAAAAL17801-06
3-Iodobenzylamine 97%
3-Iodobenzylamine

Purity: 97% CAS No.: 696-40-2 Molecular Formula: C7H8IN Formula Weight: 233.05 Flash Point: 110°(230°F) Refractive Index: 1.6375 Boiling Point: 132°/8mm Beilstein Reference: 2689602 UN No.: 2735 MDL No.: MFCD00192227

Formula: C₇H₈IN
Boiling Pt: 132 °C/8 mm
Density: 1.748
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192227
CAS Number: 696-40-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR