5-Fluoro-2-iodoaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

L17723-03 L17723-06
CAAAAL17723-03EA 95.59 CAD
CAAAAL17723-03 CAAAAL17723-06
5-Fluoro-2-iodoaniline 97%
5-Fluoro-2-iodoaniline

Purity: 97% CAS No.: 255724-71-1 Molecular Formula: C6H5FIN Formula Weight: 237.02 Melting Point: 40-42° Beilstein Reference: 8478850 UN No.: 2811 MDL No.: MFCD02093953

Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 259
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD02093953
CAS Number: 255724-71-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR