6-Bromoindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17703-03
L17703-MD
CAAAAL17703-03EA
301.78
CAD
CAAAAL17703-03
CAAAAL17703-MD
6-Bromoindole 98%
6-Bromoindole
Purity: 98%
CAS No.: 52415-29-9
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Melting Point: 91-93°
Boiling Point: 70-75°/0.01mm
MDL No.: MFCD00238550
Formula:
C₈H₆BrN Boiling Pt: 70…75 °C/0.01 mm Melting Pt: 91…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00238550 CAS Number: 52415-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...