4-Phenoxybenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Phenoxybenzene-1-sulfonyl chloride, 4-Phenoxybenzene-1-sulphonyl chloride
L17664-03
L17664-06
CAAAAL17664-03EA
97.8
CAD
CAAAAL17664-03
CAAAAL17664-06
4-Phenoxybenzenesulfonyl chloride 97%
4-Phenoxybenzenesulphonyl chloride
Purity: 97%
CAS No.: 1623-92-3
Molecular Formula: C12H9ClO3S
Formula Weight: 268.72
Melting Point: 42-45°
Boiling Point: 180°/5mm
UN No.: 3260
MDL No.: MFCD00625748
Formula:
C₁₂H₉ClO₃S Boiling Pt: 180 °C/5 mm Melting Pt: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00625748 CAS Number: 1623-92-3 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...