Potassium(4-fluorophenyl)trifluoroborate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17655-03
L17655-06
CAAAAL17655-03EA
94.64
CAD
CAAAAL17655-03
CAAAAL17655-06
Potassium(4-fluorophenyl)trifluoroborate 95%
Potassium (4-fluorophenyl)trifluoroborate
Purity: 95%
CAS No.: 192863-35-7
Molecular Formula: C6H4BF4K
Formula Weight: 202.00
Melting Point: 290-292°
Beilstein Reference: 7783544
Formula:
C₆H₄BF₄K Melting Pt: 290…292 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318170 CAS Number: 192863-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...