7-Chloroindole 98%

Supplier: Thermo Scientific Chemicals
Warning

L17637-03 L17637-MD
CAAAAL17637-03EA 154.52 CAD
CAAAAL17637-03 CAAAAL17637-MD
7-Chloroindole 98%
7-Chloroindole

Purity: 98% CAS No.: 53924-05-3 Molecular Formula: C8H6ClN Formula Weight: 151.60 Melting Point: 57-58° Beilstein Reference: 114743

Formula: C₈H₆ClN
MW: 151.6 g/mol
Melting Pt: 57…58 °C
Storage Temperature: Ambient
MDL Number: MFCD01321301
CAS Number: 53924-05-3
EINECS: 258-865-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR