7-Fluoroindole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17621-03
L17621-MD
CAAAAL17621-03EA
91.17
CAD
CAAAAL17621-03
CAAAAL17621-MD
7-Fluoroindole 97%
7-Fluoroindole
Purity: 97%
CAS No.: 387-44-0
Molecular Formula: C8H6FN
Formula Weight: 135.14
Melting Point: 61-63°
Beilstein Reference: 114740
Formula:
C₈H₆FN Melting Pt: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074502 CAS Number: 387-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...