Potassium (4-methylphenyl)trifluoroborate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium trifluoro(p-tolyl)borate, Potassium p-tolyltrifluoroborate, (4-Methylphenyl)trifluoroboric acid potassium salt

L17604-03 L17604-06
CAAAAL17604-03EA 65.41 CAD
CAAAAL17604-03 CAAAAL17604-06
Potassium (4-methylphenyl)trifluoroborate 98%
Potassium (4-methylphenyl)trifluoroborate

Purity: 98% CAS No.: 216434-82-1 Molecular Formula: C7H7BF3K Formula Weight: 198.04 Melting Point: 300° Beilstein Reference: 8290098

Formula: C₇H₇BF₃K
MW: 198.04 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318171
CAS Number: 216434-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR