Potassium phenyltrifluoroborate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17568-03
L17568-06
CAAAAL17568-03EA
59.09
CAD
CAAAAL17568-03
CAAAAL17568-06
Potassium phenyltrifluoroborate 98%
Potassium phenyltrifluoroborate
Purity: 98%
CAS No.: 153766-81-5
Molecular Formula: C6H5BF3K
Formula Weight: 184.00
Melting Point: 296-301°
Beilstein Reference: 7782070
Formula:
C₆H₅BF₃K Melting Pt: 296…301 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318172 CAS Number: 153766-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...